Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Profilin 1 Protein
SDP

Catalog No. NBP130215 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130215 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130215 Supplier R&D Systems™ Supplier No. NBP130215

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-140 of Human Profilin 1 The Recombinant Human Profilin 1 Protein is derived from E. coli. The Recombinant Human Profilin 1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number P07737
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 10% glycerol
Gene ID (Entrez) 5216
Molecular Weight (g/mol) 15kDa
Name Profilin 1 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen Profilin 1, 1-140 aa, Human, Recombinant, E.coli (NP_005013).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Product Line Novus
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.