Learn More
R&D Systems™ Recombinant Human PRPS1 His-tag Protein
Description
Specifications
Specifications
| Accession Number | P60891.2 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl, Glycerol and TCEP. |
| Gene ID (Entrez) | 5631 |
| Molecular Weight (g/mol) | M.W.-observed: 36 kDa, M.W.-theroretical: 36 kDa |
| Quantity | 50 μg |
| Source | E. coli-derived human PRPS1 protein Pro2-Leu318 with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CMTX5, CMTX5ARTS, deafness, X-linked 2, perceptive, congenital, DFN2, DFNX1, DFNX1KIAA0967, dJ1070B1.2 (phosphoribosyl pyrophosphate synthetase 1), EC 2.7.6.1, Phosphoribosyl pyrophosphate synthase I, phosphoribosyl pyrophosphate synthetase 1, PPRibP, PRS I, PRSI, PRS-I, ribose-phosphate diphosphokinase 1, ribose-phosphate pyrophosphokinase 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.