Learn More
R&D Systems™ Recombinant Human PSG6 Protein
Description
Order Info
Ships at ambient temperature.
Specifications
Specifications
| Accession Number | Q00889-2 |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS and NaCl with Trehalose |
| Gene ID (Entrez) | 3730 |
| Molecular Weight (g/mol) | 45 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell,HEK293-derived human PSG6 protein Gln35-His424,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method |
| Biological Activity | 0.9 to 4.5μg/mL |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.