Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human PSMA/FOLH1/NAALADase I His Avi Protein

Catalog No. AVI4234025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AVI4234025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AVI4234025 Supplier R&D Systems™ Supplier No. AVI4234025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Biotinylated

Measured by its binding ability in a functional ELISA. When Human PSMA/FOLH1 Affinity Purified Polyclonal Antibody (Catalog # AF4234 ) is immobilized at 2 μg/mL, 100 μL/well, it binds Biotinylated Recombinant Human PSMA/FOLH1/NAALADase I His-tag Avi-tag with an ED50 of 0.01-0.08 μg/mL. Measured by its ability to hydrolyze the substrate N-acetyl-L-Asp-L-Glu into N-acetyl-L-Asp and L-Glu. The L-Glu product is measured by fluorescence after its derivatization by ortho-phthaldialdehyde. The specific activity is > 250 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number Q04609.1
For Use With (Application) Bioactivity, Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in MES and NaCl.
Gene ID (Entrez) 2346
Molecular Weight (g/mol) M.W.-observed: 94-107 kDa, under reducing conditions, M.W.-theoretical: 83 kDa
Quantity 25 μg
Source Chinese Hamster Ovary cell line, CHO-derived human PSMA/FOLH1/NAALADase I protein Avi-tag HHHHHH Human PSMA/FOLH1/NAALADase I (Lys44-Ala750) (N-terminus) Accession # Q04609.1 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias Cell growth-inhibiting gene 27 protein, cell growth-inhibiting protein 27, EC 3.4.17.21, FGCP, folate hydrolase (prostate-specific membrane antigen) 1, Folate hydrolase 1, FOLH1, FOLHNAALADase I, Folylpoly-gamma-glutamate carboxypeptidase, GCP2, GCPII, GCPIImGCP, glutamate carboxylase II, glutamate carboxypeptidase 2, Glutamate carboxypeptidase II, Membrane glutamate carboxypeptidase, mopsm, NAALAD1, NAALAD1N-acetylated-alpha-linked acidic dipeptidase I, NAALAdase, NAALADase I, N-acetylated alpha-linked acidic dipeptidase 1, prostate specific membrane antigen variant F, Prostate-specific membrane antigen, PSMAFGCP, PSMPteroylpoly-gamma-glutamate carboxypeptidase
Conjugate Biotin
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PSMA/FOLH1/NAALADase I
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.