Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human PTEN Protein

Catalog No. 847PN010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
847PN010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 847PN010 Supplier R&D Systems™ Supplier No. 847PN010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Inhibition Activity

Measured by its ability to dephosphorylate either diC8PIP3 or diC16PIP3. The specific activity is >480 nmol/min/mg, as measured under the described conditions.

Specifications

Accession Number P60484
For Use With (Application) Inhibition Assay
Formulation Supplied as a 0.2 μm filtered solution in HEPES, NaCl, EDTA, Glycerol and Betamercaptoethanol.
Gene ID (Entrez) 5728
Molecular Weight (g/mol) M.W. (Observed): 59 kDa, reducing conditions; M.W. (theoretical): 48 kDa
Name PTEN
Quantity 10 μg
Source E. coli-derived human PTEN protein Thr2-Val403, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol PTEN
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >80%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PTEN
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.