Learn More
Description
Specifications
Specifications
| For Use With (Application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulation | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gene ID (Entrez) | 7179 |
| Molecular Weight (g/mol) | 36.74 kDa |
| Quantity | 25 μg |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Gene Alias | Cancer/testis antigen 44, EC 3.1.3.48, PTEN2, putative tyrosine-protein phosphatase TPTE, tensin, putative protein-tyrosine phosphatase, transmembrane phosphatase with tensin homologytensin, putative protein-tyrosine phosphatase, EC 3.1.3.4810CT44PTEN-related tyrosine phosphatase, Tumor antigen BJ-HCC-5 |
| Gene Symbol | TPTE |
| Product Line | Novus |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
