Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human PTPRD Fc Chimera Protein

Catalog No. 9995PR050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
99-95P-R050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 99-95P-R050 Supplier R&D Systems™ Supplier No. 9995PR050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its binding ability in a functional ELISA. When Recombinant Human PTPRD Fc Chimer is immobilized at 1 μg/mL, 100 μL/well, Recombinant Human IL-1 RAPL1/IL-1 R8 Fc Chimera (Catalog # 9949-ML ) binds with an ED50 of 1.5-9 ng/mL.

Specifications

Accession Number P23468
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 5789
Molecular Weight (g/mol) M.W. (Observed): 137-151 kDa, reducing conditions; M.W. (theoretical): 154 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human PTPRD protein Human PTPRD (Glu21-Ser1174) (N-terminus) Accession # P23468 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Specific Reactivity PTPRD
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 400 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Purity or Quality Grade >80%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PTPRD
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.