Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 25797 |
| Species | Human |
| Product Line | R&D Systems |
| Purity | >95% by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
| Accession Number | Q16769 |
| Content And Storage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol. |
| Gene Alias | EC 2.3.2.5, GCT, Glutaminyl cyclase, glutaminyl-peptide cyclotransferase, Glutaminyl-tRNA cyclotransferase, Glutamyl cyclase, QCEC, sQC |
| Gene Symbol | QPCT |
| Molecular Weight (g/mol) | M.W. (Observed): 36-40 kDa, reducing conditions; M.W. (theoretical): 38 kDa |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
