Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human QSOX1/Quiescin Q6 Protein

Catalog No. 9209QS020
Change view
Click to view available options
Recombinant Human QSOX1/Quiescin Q6 Protein, Carrier-Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
92-09Q-S020 Recombinant Human QSOX1/Quiescin Q6 Protein, Carrier-Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 92-09Q-S020 Supplier R&D Systems™ Supplier No. 9209QS020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human QSOX1/Quiescin Q6 Protein is derived from E. coli. The Recombinant Human QSOX1/Quiescin Q6 Protein has been validated for the following applications: Enzyme Activity.

Order Info

The product is shipped at ambient temperature.

Specifications

For Use With (Application) Enzyme Activity
Source E. coli-derived human QSOX1/Quiescin Q6 protein Ser33-Ala546,with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Conjugate Unconjugated

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.