Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human R-Spondin 1 Biotinylated Protein
SDP

Catalog No. BT4645025 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT4645025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT4645025 Supplier R&D Systems™ Supplier No. BT4645025

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human R-Spondin 1 Biotinylated Protein is derived from CHO. The Recombinant Human R-Spondin 1 Biotinylated Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q2MKA7
For Use With (Application) Bioactivity
Gene ID (Entrez) 284654
Molecular Weight (g/mol) Theoretical M.W.: 26kDa (unlabeled)
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived human R-Spondin 1 protein Arg31-Ala263
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Cristin 3, CRISTIN3, FLJ40906Roof plate-specific spondin-1, hRspo1, roof plate-specific spondin, RSPO, RSPO1, RSPONDIN, RSpondin 1, R-spondin homolog (Xenopus laevis), R-spondin1, R-spondin-1
Common Name R-Spondin 1
Gene Symbol RSPO1
Biological Activity Measured by its ability to induce Topflash reporter activity in HEK293T human embryonic kidney cells. The ED50 for this effect is typically 5-30ng/mL in the presence of 5ng/mL recombinant mouse Wnt-3a.
Conjugate Biotin
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Arg31-Ala263
Product Line R&D Systems
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.