Learn More
Description
Specifications
Specifications
| Accession Number | NP_000955 |
| For Use With (Application) | Western Blot |
| Formulation | 20mM Tris buffer (pH 7.5), 0.1 M NaCl, 5mM beta-mercaptoethanol |
| Gene ID (Entrez) | 5914 |
| Name | RAR alpha/NR1B1 Protein |
| Purification Method | Protein |
| Immunogen | MGSSHHHHHH SSGLVPRGSH MSEEIVPSPP SPPPLPRIYK PCFVCQDKSS GYHYGVSACE GCKGFFRRSI QKNMVYTCHR DKNCIINKVT RNRCQYCRLQ KCFEVGMSKE SVRNDRNKKK KEVPKPE |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
