Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human RBBP4/RbAp48 Protein
SDP

Catalog No. NB171825 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171825 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171825 Supplier Novus Biologicals™ Supplier No. NBP318203
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA. Immobilized Human RBBP4/RbAp48 Protein at 1 ug/mL (100 ul/well) can bind RBBP4/RbAp48 Rabbit pAb with a linear range of 0.976-7.09 ng/mL.

Specifications

Concentration LYOPH
For Use With (Application) Bioactivity, SDS-PAGE
Formulation Lyophilized from a 0.22 um filtered solution of 50mM Tris, 500mM NaCl, pH 7.4
Gene ID (Entrez) 5928
Quantity 50 μg
Storage Requirements Store at -20°C.
Endotoxin Concentration <1.0 EU/μg of the protein by LAL method
Gene Alias CAF-1 subunit C, CAF-I p48, Chromatin assembly factor 1 subunit C, Chromatin assembly factor I p48 subunit, chromatin assembly factor/CAF-1 p48 subunit, histone-binding protein RBBP4, MSI1 protein homolog, Nucleosome-remodeling factor subunit RBAP48, NURF55, RbAp48, RBBP-4, retinoblastoma binding protein 4, Retinoblastoma-binding protein 4CAF-I 48 kDa subunit, Retinoblastoma-binding protein p48
Gene Symbol RBBP4
Reconstitution Reconstitute to a concentration of 0.1-0.5 mg/mL in sterile distilled water
Research Category Cancer, Cell Cycle and Replication, Chromatin Research, Growth and Development, Stem Cell Markers, Tumor Suppressors
Purity or Quality Grade >92%, by SDS-PAGE
Protein RBBP4/RbAp48
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.