Learn More
R&D Systems™ Recombinant Human RECK His-tag Protein
Description
Specifications
Specifications
| Accession Number | O95980 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 8434 |
| Molecular Weight (g/mol) | Observed: 103 - 122 kDa, under reducing conditions; Theroretical: 102 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human RECK protein Gly27-Pro941, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 250 μg/mL aq. soln.. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.