Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Reelin Protein

Catalog No. 8546MR050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-46M-R050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 85-46M-R050 Supplier R&D Systems™ Supplier No. 8546MR050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Reelin Protein is derived from CHO. The Recombinant Human Reelin Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P78509
For Use With (Application) Bioactivity
Gene ID (Entrez) 5649
Molecular Weight (g/mol) Observed M.W.: 155-188kDa, reducing conditions, Theoretical M.W.: 163kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Reelin protein Ser1221-Gln2666,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias EC 3.4.21, EC 3.4.21.-, Reeler, reelin, RELN, RL, RLPRO1598
Common Name Reelin
Gene Symbol RELN
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human Reelin is coated at 1.5μg/mL (100μL/well), the concentration of Recombinant Human Apolipoprotein E R2/ApoE R2 that produces 50% optimal binding response is typically 50-300ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Ser1221-Gln2666, with a C-terminal 6-His tag
Purity or Quality Grade >90%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.