Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Relaxin-2 (B-29/A-24) Protein

Catalog No. 6586RN025CF
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Sodium Acetate.
Lyophilized from a 0.2μm filtered solution in sodium acetate with BSA as a carrier protein.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
6586RN025CF Lyophilized from a 0.2μm filtered solution in Sodium Acetate.
6586RN025 Lyophilized from a 0.2μm filtered solution in sodium acetate with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 6586RN025CF Supplier R&D Systems™ Supplier No. 6586RN025/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Relaxin-2 (B-29/A-24) Protein is derived from E. coli. The Recombinant Human Relaxin-2 (B-29/A-24) Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Sodium Acetate.
Gene ID (Entrez) 6019
Molecular Weight (g/mol) 2.7 kDa (A chain); 3.3 kDa (B chain)
Name Relaxin-2 (B-29/A-24)
Quantity 25 μg
Source E. coli-derived human Relaxin-2 protein Asp25-Ser53 (B chain) & Gln162-Cys185 (A chain)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked heterodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.