Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Ret His-tag Protein

Catalog No. 10302CR050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-302-CR050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-302-CR050 Supplier R&D Systems™ Supplier No. 10302CR050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its binding ability in a functional ELISA.

When Recombinant Human Ret His-tag is immobilized at 1 μg/mL (100 μL/well), Recombinant Human GDNF binds with an ED50 of 0.03 to 0.3 μg/mL in the presence of Recombinant Human GFR alpha -1/GDNF R alpha-1 Fc Chimera

Specifications

Accession Number P07949
For Use With (Application) Bioactivity
Gene ID (Entrez) 5979
Molecular Weight (g/mol) 69 kDa
Name CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human Ret protein Leu29-Arg635, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.