Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human RPL11 His Protein
SDP

Catalog No. NBP223417 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223417 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP223417 Supplier R&D Systems™ Supplier No. NBP223417

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-178 of Human RPL11 The Recombinant Human RPL11 Protein is derived from E. coli. The Recombinant Human RPL11 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_000966
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.2 M NaCl, 50% glycerol, 1mM DTT, 2mM EDTA, 250mM Imidazole
Gene ID (Entrez) 6135
Molecular Weight (g/mol) 22.6kDa
Name RPL11 Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen RPL11, 1-178aa, Human, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MGSMAQDQGE KENPMRELRI RKLCLNICVG ESGDRLTRAA KVLEQLTGQT PVFSKARYTV RSFGIRRNEK IAVHCTVRGA KAEEILEKGL KVREYELRKN NFSDTGNFGF GIQEHIDLGI KYDPSIGIYG LDFYVVLGRP GFSIADKKRR TGCIGAKHRI SKEEAMRWFQ QKYDGIILPG K
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category DNA replication Transcription Translation and Splicing
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.