Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human RXR alpha/NR2B1 Protein
SDP

Catalog No. NBC118416 Shop All R&D Systems Products
Change view
Click to view available options
0.1mg; Unlabeled
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBC118416 0.1mg; Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118416 Supplier Novus Biologicals™ Supplier No. NBC118416
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids111-228 of Human RXR alpha/NR2B1 The Recombinant Human RXR alpha/NR2B1 Protein is derived from E. coli. The Recombinant Human RXR alpha/NR2B1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_002948
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 7.5) 0.1 M NaCl, 5mM beta-Mercaptoethanol
Gene ID (Entrez) 6256
Molecular Weight (g/mol) theoretical_molecular_weight: L606
Name RXR alpha/NR2B1 Protein
Purification Method Protein
Immunogen MLGLNGVLKVPAHPSGNMASFTKHICAICGDRSSGKHYGVYSCEGCKGFFKRTVRKDLTYTCRDNKDCLIDKRQRNRCQYCRYQKCLAMGMKREAVQEERQRGKDRNENEVESTSSANE
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.