Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human S100A11 Protein

Catalog No. 9015S11050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-15S-11050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-15S-11050 Supplier R&D Systems™ Supplier No. 9015S11050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human S100A11 Protein is derived from E. coli. The Recombinant Human S100A11 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P31949
For Use With (Application) Bioactivity
Gene ID (Entrez) 6282
Molecular Weight (g/mol) Observed M.W.: 9kDa, reducing conditions, Theoretical M.W.: 12kDa
Quantity 50 μg
Source E. coli-derived human S100A11 protein Ala2-Thr105
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias calgizzarin, Metastatic lymph node gene 70 protein, MLN 70, MLN70, protein S100-A11, Protein S100-C, S100 calcium binding protein A11, S100 calcium-binding protein A11, S100 calcium-binding protein A11 (calgizzarin), S100C, S100CS100 calcium binding protein A11 (calgizzarin)
Common Name S100A11
Gene Symbol S100A11
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human ANXA-2 is immobilized at 0.5μg/mL (100μL/well), the concentration of Recombinant Human S100A11 that produces 50% of the optimal binding response is approximately 0.4-2.4μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived Ala2-Thr105
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.