Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human S100A8 Protein
SDP

Catalog No. 9876S8050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
98-76S-8050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 98-76S-8050 Supplier R&D Systems™ Supplier No. 9876S8050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human S100A8 Protein is derived from E. coli. The Recombinant Human S100A8 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P05109
For Use With (Application) Bioactivity
Gene ID (Entrez) 6279
Molecular Weight (g/mol) 11 kDa
Quantity 50 μg
Source E. coli-derived human S100A8 protein Met1-Glu93
Storage Requirements 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Specific Reactivity S100A8
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 150 μg/mL in water.
Buffer Lyophilized from a 0.2 μm filtered solution in Tris and TCEP with Trehalose.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.