Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human SCF Protein
GSA_VASDP

Catalog No. 255SC001MGC Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
200 μg
50 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
255SC001MGC 1 mg Lyophilized from a 0.2μm filtered solution in PBS.
255SC010 10 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
255SC010CF 10 μg Lyophilized from a 0.2μm filtered solution in PBS.
255SC050 50 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
255SC050CF 50 μg Lyophilized from a 0.2μm filtered solution in PBS.
25-5SC-200 200 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
25-5SC-200CF 200 μg Lyophilized from a 0.2μm filtered solution in PBS.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 255SC001MGC Supplier R&D Systems™ Supplier No. 255SC001MG/CF

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 1-5 ng/mL.

Order Info

Ships at ambient temperature.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 4254
Molecular Weight (g/mol) M.W. (theroretical): 19 kDa
Name SCF/c-kit Ligand
Quantity 1 mg
Source E. coli-derived human SCF/c-kit Ligand protein Glu26-Ala189,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor
Conjugate Unconjugated
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.