Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human SCF Protein

Catalog No. 11010SC010
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11010SC010 10 μg
11010SC01M 1 mg
11010SC050 50 μg
11010SC100 100 μg
11010SC500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 11010SC010 Supplier R&D Systems™ Supplier No. 11010SC010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 1.00-5.00 ng/mL.

Specifications

Accession Number P21583.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 4254
Molecular Weight (g/mol) MolecularWeight-observed: 22-31 kDa, under reducing conditions.; MolecularWeight-theroretical: 18 kDa
Quantity 10 μg
Source Human embryonic kidney cell, HEK293-derived human SCF/c-kit Ligand protein Glu26-Ala189
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor
Conjugate Unconjugated
Reconstitution Reconstitute at 100-500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein SCF/c-kit Ligand
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.