Learn More
Description
Specifications
Specifications
| For Use With (Application) | Western Blot, ELISA, SDS-PAGE, Protein Array, Immunoaffinity Purification |
| Formulation | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gene ID (Entrez) | 8436 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 73.6 kDa |
| Quantity | 25 μg |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Gene Alias | cavin-2, Phosphatidylserine-binding protein, PS-p68serum deprivation response (phosphatidylserine-binding protein), SDRphosphatidylserine binding protein, serum deprivation response, serum deprivation response (phosphatidylserine binding protein), serum deprivation-response protein |
| Gene Symbol | SDPR |
| Product Line | Novus |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
