Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Semaphorin 3B Fc Chimera Protein

Catalog No. 9518S3025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9518S3025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 9518S3025 Supplier R&D Systems™ Supplier No. 9518S3025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Semaphorin 3B Fc Chimera Protein is derived from T. ni (baculovirus). The Recombinant Human Semaphorin 3B Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q13214-1
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in Citric Acid, NaCl and Trehalose.
Gene ID (Entrez) 7869
Molecular Weight (g/mol) 105kDa
Quantity 25 μg
Source Trichoplusia ni,T. ni (baculovirus)-derived human Semaphorin 3B protein, (N-terminus) Human Semaphorin 3B (Ala25-Gly727(Arg551Ala,Arg554Ala)) Accession &Num; Q13214-1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Gene Alias (semaphorin) 3B, LUCA-1, SemA, Sema A(V), sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema V, SEMA3B, sema5, SEMA5FLJ34863, SEMAA, semaphorin A, semaphorin V, semaphorin-3B, Semaphorin-V, SEMAV
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500μg/mL in water.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.