Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Semaphorin 3E Protein

Catalog No. 3239S3B025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-39S-3B025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 32-39S-3B025 Supplier R&D Systems™ Supplier No. 3239S3B025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Semaphorin 3E Protein is derived from NS0. The Recombinant Human Semaphorin 3E Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number O15041
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS, NaCl and Tween™ 20 with Trehalose.
Gene ID (Entrez) 9723
Molecular Weight (g/mol) Observed M.W.: 85-100kDa, 66kDa and 25kDa, reducing conditions, Theoretical M.W.: 88kDa
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived human Semaphorin 3E protein His24-Ser775 (Arg557Ala and Arg560Ala),with an N-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Gene Alias (semaphorin) 3E, coll-5, KIAA0331M-SEMAH, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema3E, SEMAH, SEMAHM-SemaK, semaphorin-3E
Common Name Semaphorin 3E
Gene Symbol SEMA3E
Biological Activity Measured by its ability to inhibit the proliferation of HUVEC human umbilical vein endothelial cells. Moriya, J. et al. (2010) Circ. Res. 106:391. The ED50 for this effect is typically 0.04-0.24μg/mL. Measured by its binding ability in a functional ELISA. When Recombinant Human Plexin D1 is coated at 5μg/mL, Recombinant Human Semaphorin 3E binds with an apparent Kd <0.4 nM.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived His24-Ser775 (Arg557Ala and Arg560Ala), with an N-terminal 10-His tag
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.