Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Semaphorin 4F Fc Chimera Protein

Catalog No. 10113S4050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-113-S4050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-113-S4050 Supplier R&D Systems™ Supplier No. 10113S4050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its ability to enhance neurite outgrowth of E16-E18 rat embryonic cortical neurons.

Recombinant Human Semaphorin 4F Fc Chimera immobilized at 2.5 μg/mL on a 96 well plate and is able to significantly enhance neurite outgrowth

Specifications

Accession Number O95754-1
For Use With (Application) Bioactivity
Gene ID (Entrez) 10505
Molecular Weight (g/mol) 96 to 111 kDa, under reducing conditions
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Semaphorin 4F protein. Human Semaphorin 4F (Arg35-Val659) Accession # O95754-1IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration < 0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 250 μg/mL in Water
Buffer Lyophilized from a 0.2 μm filtered solution in Citric Acid and NaCl
Purity or Quality Grade > 85%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.