Learn More
R&D Systems™ Recombinant Human Serpin A12 Protein
Description
Specifications
Specifications
| Accession Number | Q8IW75 |
| Formulation | Supplied as a 0.2μm filtered solution in Tris and NaCl |
| Gene ID (Entrez) | 145264 |
| Molecular Weight (g/mol) | 47kDa |
| Name | Serpin A12 |
| Quality Control Testing | SDS-PAGE: 54-60kDa, reducing conditions |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human Serpin A12 protein Leu20-Lys414,with C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.