Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Serpin B2/PAI-2 Protein
SDP

Catalog No. 9206PI025 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
92-06P-I025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 92-06P-I025 Supplier R&D Systems™ Supplier No. 9206PI025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Serpin B2/PAI-2 Protein is derived from E. coli. The Recombinant Human Serpin B2/PAI-2 Protein has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number P05120
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 5055
Molecular Weight (g/mol) Observed M.W.: 41kDa, reducing conditions, Theoretical M.W.: 44kDa
Quantity 25 μg
Source E. coli-derived human Serpin B2 protein Glu2-Ala65/Ala99-Pro415,with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Gene Alias HsT1201, Monocyte Arg-serpin, PAI, PAI2, PAI2Urokinase inhibitor, Placental plasminogen activator inhibitor, PLANH2, PLANH2PAI-2, plasminogen activator inhibitor 2, plasminogen activator inhibitor, type II (arginine-serpin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 2, Serpin B2, serpin peptidase inhibitor, clade B (ovalbumin), member 2, Urokinase Inhibitor
Common Name SerpinB2
Biological Activity Measured by its ability to inhibit uPA cleavage of a peptide substrate, N-carbobenzyloxy-Gly-Gly-Arg-7-amido-4-methylcoumarin (Z-GGR-AMC). The inhibition IC50 is <15 nM, as measured under the described conditions. See Activity Assay Protocol on RnDSystems.com.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived Glu2-Ala65/Ala99-Pro415, with an N-terminal Met and 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.