Learn More
Description
Order Info
Ships at ambient temperature.
Specifications
Specifications
| Accession Number | P48594 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Lyophilized from a 0.2μ filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 6318 |
| Molecular Weight (g/mol) | 46 kDa |
| Quantity | 50 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Serpin B4 protein Asn2-Pro390 with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
