Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human SETD7/9 Protein
SDP

Catalog No. NBC118444 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118444 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118444 Supplier Novus Biologicals™ Supplier No. NBC118444
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-366 of Human SETD7/9 The Recombinant Human SETD7/9 Protein is derived from E. coli. The Recombinant Human SETD7/9 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_085151
For Use With (Application) ELISA, SDS-PAGE
Formulation 50mM Tris-HCl buffer (pH7.5) 0.2 M NaCl, 5mM DTT, 20% glycerol
Gene ID (Entrez) 80854
Molecular Weight (g/mol) 40.7kDa
Name SETD7/9 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen Human, Recombinant, E.coli. 40.7kDa (366 aa), confirmed by MALDI-TOF
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Research Category Histones and Modified Histones, Signal Transduction
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.