Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human SHBG Protein

Catalog No. 9614SG050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
96-14S-G050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 96-14S-G050 Supplier R&D Systems™ Supplier No. 9614SG050
Add to Cart
Add to Cart
This item is not returnable. View return policy

R&D Systems™ Recombinant Human SHBG Protein CF is a variably glycosylated, secreted non-disulfide linked homodimer that belongs to the SHBG family.

The Recombinant Human SHBG Protein is derived from NS0.

Specifications

Accession Number P04278-1
For Use With (Application) Flow Cytometry
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 6462
Molecular Weight (g/mol) 41 kDa
Name ABP, ABPTestosterone-estrogen-binding globulin, androgen binding protein, MGC126834, MGC138391, SBP, sex hormone-binding globulin, Sex steroid-binding protein, TeBG, Testis-specific androgen-binding protein, testosterone-binding beta-globulin, Testosterone-estradiol-binding globulin
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human SHBG protein Leu30-His402,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 250μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.