Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Siglec-3/CD33 Fc Alexa Fluor™ 647 Protein

Catalog No. AFR1137020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AFR1137020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AFR1137020 Supplier R&D Systems™ Supplier No. AFR1137020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured by flow cytometry for its ability to bind anti-human Siglec-3/CD33 Monoclonal Antibody conjugated beads.The concentration of Recombinant Human Siglec-3/CD33 Fc Chimera Alexa Fluor™ 647 (Catalog # AFR1137) that produces 50% of the binding response is 0.50-20.0 ng/mL.

Specifications

Accession Number AAA51948
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 945
Molecular Weight (g/mol) M.W.-observed: 67-85 kDa, under reducing conditions, M.W.-theroretical: 53.4 kDa (monomer)
Quantity 20 μg
Source Mouse myeloma cell line, NS0-derived human Siglec-3/CD33 protein Human Siglec-3 (Asp18-His259)(Val257Leu) (N-terminus) Accession # AAA51948 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias CD33, CD33 antigen, CD33 antigen (gp67), CD33 molecule, FLJ00391, gp67, myeloid cell surface antigen CD33, p67, sialic acid binding Ig-like lectin 3, Sialic acid-binding Ig-like lectin 3, Siglec3, SIGLEC-3, SIGLEC3gp67
Conjugate Alexa Fluor 647
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Siglec-3/CD33
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.