Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Siglec-8 Fc Chimera Protein

Catalog No. 9045SL050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-45S-L050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-45S-L050 Supplier R&D Systems™ Supplier No. 9045SL050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Siglec-8 Fc Chimera Protein is derived from NS0. The Recombinant Human Siglec-8 Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number AAH53319
For Use With (Application) Bioactivity
Gene ID (Entrez) 27181
Molecular Weight (g/mol) Observed M.W.: 68-88kDa, reducing conditions, Theoretical M.W.: 64kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human Siglec-8 protein, (N-terminus) Human Siglec-8 (Met17-Ala363) Accession &Num; AAH53319 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CD329 antigen, CDw329, MGC59785, SAF2, SAF2SAF-2, sialic acid binding Ig-like lectin 8, Sialoadhesin family member 2, Siglec8, Siglec-8, SIGLEC8L, SIGLEC8Lsialic acid-binding Ig-like lectin 8
Common Name Siglec-8
Gene Symbol SIGLEC8
Biological Activity Measured by its binding ability in a functional ELISA. When Biotinylated Sialyl Lewis X was immobilized at 0.05μg/mL (100μL/well), it binds Recombinant Human Siglec-8 Fc Chimera. The concentration of Human Siglec-8 Fc Chimera that produces 50% of the optimal binding response is approximately 0.2-1.2μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Human Siglec-8 (Met17-Ala363) Accession # AAH53319, N-terminus; IEGRMD; Human IgG1 (Pro100-Lys330), C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.