Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human SIRP alpha/CD172a Alexa Fluor™ 488 Protein

Catalog No. AFG4546020
Change view
Click to view available options
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AFG4546020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AFG4546020 Supplier R&D Systems™ Supplier No. AFG4546020
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Fc Chimera

Measured by flow cytometry for its ability to bind Human SIRP alpha/CD172a Antibody conjugated fluorescent beads at 0.313-1.25 μg/mL (100 μL/well). Please Note: Optimal dilutions should be determined by each laboratory for each application.

Specifications

Accession Number NP_542970.1
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 140885
Molecular Weight (g/mol) MolecularWeight-observed: 70-105 kDa, under reducing conditions., MolecularWeight-theroretical: 64 kDa (monomer)
Quantity 20 μg
Source Chinese Hamster Ovary cell line, CHO-derived human SIRP alpha/CD172a protein Human SIRP alpha (Gly27-Arg370) & (Glu31-Arg370) (N-terminus) Accession # NP_542970.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias Bit, BITbrain-immunoglobulin-like molecule with tyrosine-based activation motifs, Brain Ig-like molecule with tyrosine-based activation motifs, CD172 antigen-like family member A, CD172a, CD172a antigen, Inhibitory receptor SHPS-1, Macrophage fusion receptor, MFR, MFRtyrosine phosphatase SHP substrate 1, MYD1, MYD-1, MyD-1 antigen, P84, protein tyrosine phosphatase, non-receptor type substrate 1, PTPNS1, SHP substrate 1, SHPS1, SHPS-1, SHPS1CD172A, signal-regulatory protein alpha, Signal-regulatory protein alpha-1, Signal-regulatory protein alpha-2, Signal-regulatory protein alpha-3, SIRPA, SIRPalpha, SIRP-ALPHA-1, Sirp-alpha-2, SIRPalpha2, Sirp-alpha-3, SIRPtyrosine-protein phosphatase non-receptor type substrate 1
Conjugate Alexa Fluor 488
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein SIRP alpha/CD172a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.