Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human SIRP alpha/CD172a Fc Chimera Protein

Catalog No. 4546SA050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4546SA050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 4546SA050 Supplier R&D Systems™ Supplier No. 4546SA050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by the ability of the immobilized protein to support the adhesion of human red blood cells. Subramanian, S. et al. (2006) Blood 107:2548. Red blood cells, added at 10 x 106 cells/mL (100 μL/well) to SIRP alpha coated plates, will adhere after 1 hour at 37° C. The ED50 for this effect is 0.6-2.4 μg/mL. Optimal dilutions should be determined by each laboratory for each application.

Specifications

Accession Number NP_542970
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 140885
Molecular Weight (g/mol) M.W. (Observed): 70-105 kDa, reducing conditions; M.W. (theoretical): 64 kDa (monomer)
Name SIRP alpha/CD172a Fc Chimera
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human SIRP alpha/CD172a protein Human SIRP alpha (Gly27-Arg370) & (Glu31-Arg370) (N-terminus) Accession # NP_542970 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol SIRPA
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein SIRP alpha/CD172a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.