Learn More
R&D Systems™ Recombinant Human SKP1/SKP2 Complex Protein, CF
Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | X mg/mL (X μM) in 50 mM HEPES pH 7.5, 150 mM NaCl, 10% (v/v) Glycerol, 1 mM TCEP |
| Molecular Weight (g/mol) | M.W. (theoretical): 19 kDa (SKP1), 48 kDa (SKP2) |
| Purification Method | >85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain |
| Quantity | 25 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived human SKP1/SKP2 Complex protein |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 36 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method |
| Conjugate | Unconjugated |
| Buffer | X mg/mL (X μM) in 50 mM HEPES pH 7.5, 150 mM NaCl, 10% (v/v) Glycerol, 1 mM TCEP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.