Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human SLC26A1 Protein
SDP

Catalog No. NB816443 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB816443 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB816443 Supplier Novus Biologicals™ Supplier No. H00010861G0110UG
Add to Cart
Add to Cart

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.

Specifications

For Use With (Application) Immunoaffinity Purification
Formulation 25 mM Tris-HCl of pH8.0 containing 2% glycerol.
Gene ID (Entrez) 10861
Molecular Weight (g/mol) 77.2 kDa
Quantity 10 μg
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Gene Alias EDM4, SAT1, SAT-1sulfate anion transporter 1, solute carrier family 26 (sulfate transporter), member 1, Solute carrier family 26 member 1, sulfate anion tranporter AT1, sulfate transporter, sulfate/anion transporter SAT-1 protein
Gene Symbol SLC26A1
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein SLC26A1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.