Learn More
Description
Specifications
Specifications
| For Use With (Application) | Western Blot, ELISA, PAGE, Protein Array, Immunoaffinity Purification |
| Formulation | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gene ID (Entrez) | 54407 |
| Molecular Weight (g/mol) | 31.46 kDa |
| Quantity | 10 μg |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Gene Alias | Amino acid transporter A2, ata2, ATA2System A transporter 1, KIAA1382System N amino acid transporter 2, pro1068, Protein 40-9-1, sat2, SAT2PRO1068, snat2, SNAT2amino acid transporter 2, sodium-coupled neutral amino acid transporter 2, Solute carrier family 38 member 2, solute carrier family 38, member 2, System A amino acid transporter 2 |
| Gene Symbol | SLC38A2 |
| Product Line | Novus |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
