Learn More
R&D Systems™ Recombinant Human Slit1 Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in MOPS and NaCl. |
| Gene ID (Entrez) | 6585 |
| Molecular Weight (g/mol) | 98.8 kDa |
| Name | Slit1 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Slit1 protein Met1-Gln915,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.