Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Slit3 (aa 1120-1523) Protein

Catalog No. 9067SL050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-67S-L050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-67S-L050 Supplier R&D Systems™ Supplier No. 9067SL050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Slit3 (aa 1120-1523) Protein is derived from HEK293. The Recombinant Human Slit3 (aa 1120-1523) Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number O75094
For Use With (Application) Bioactivity
Gene ID (Entrez) 6586
Molecular Weight (g/mol) Observed M.W.: 56-63kDa, reducing conditions, Theoretical M.W.: 45kDa
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human Slit3 protein Thr1120-Ser1523,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias KIAA0814, MEGF5, MEGF5Multiple epidermal growth factor-like domains protein 5, Multiple EGF-like domains protein 5, SLIL2, SLIL2FLJ10764, slit homolog 3 (Drosophila), slit homolog 3 protein, SLIT1, slit2, Slit-3slit (Drosophila) homolog 3
Common Name Slit3
Gene Symbol SLIT3
Biological Activity Measured by its ability to enhance neurite outgrowth of E16-E18 rat embryonic cortical neurons. Recombinant Human Slit3, immobilized at 1.25-2.5μg/mL on a 96-well plate, is able to significantly enhance neurite outgrowth.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Thr1120-Ser1523, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.