Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Slit3 (aa 34-1116) His-tag Protein

Catalog No. 9255SLB050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
92-55S-LB050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 92-55S-LB050 Supplier R&D Systems™ Supplier No. 9255SLB050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Carrier Free Recombinant Protein

Measured by its ability to enhance neurite outgrowth of E16-E18 rat embryonic cortical neurons. Recombinant Human Slit3 His-tag (Catalog # 9255-SLB) enhances neurite outgrowth at 5 μg/mL.

Specifications

Accession Number NP_003053.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose.
Gene ID (Entrez) 6586
Molecular Weight (g/mol) MolecularWeight-observed: 119-149 kDa, under reducing conditions., MolecularWeight-theroretical: 121 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human Slit3 protein Cys34-Val1116, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias KIAA0814, MEGF5, MEGF5Multiple epidermal growth factor-like domains protein 5, Multiple EGF-like domains protein 5, SLIL2, SLIL2FLJ10764, slit homolog 3 (Drosophila), slit homolog 3 protein, SLIT1, slit2, Slit-3 slit (Drosophila) homolog 3
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Slit3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.