Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Sonic Hedgehog/Shh (C24II) Animal-Free

Catalog No. AFL1845025
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AFL1845025 25 μg
AFL1845100 100 μg
AFL1845500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AFL1845025 Supplier R&D Systems™ Supplier No. AFL1845025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Also available, GMP Shh Protein, Catalog # 1845-GMP! Animal-free and manufactured under GMP guidelines for therapeutic manufacturing applications.

Measured by its ability to induce alkaline phosphatase production by C3H10T1/2 mouse embryonic fibroblast cells. Nakamura, T. et al. (1997) Biochem. Biophys. Res. Commun. 237:465. The ED50 for this effect is 0.1-0.4 μg/mL.

Specifications

Accession Number NP_000184.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and NaCl.
Gene ID (Entrez) 6469
Molecular Weight (g/mol) M.W.-observed: 22 kDa, reducing conditions, M.W.-theroretical: 20 kDa
Quantity 25 μg
Source E. coli-derived human Sonic Hedgehog/Shh protein Cys24-Gly197 (Cys24Ile-Ile), with an N-terminal Met Produced using non-animal reagents in an animal-free laboratory.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Alias HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Conjugate Unconjugated
Reconstitution Reconstitute at 100-200 μg/mL in PBS, and allow up to 24 hours (at 2°C to 8°C for complete reconstitution.)
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein Sonic Hedgehog/Shh
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.