Learn More
R&D Systems™ Recombinant Human Sphingosine Kinase 2/SPHK2 Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in MES, NaCl, Glycerol and DTT. |
| Gene ID (Entrez) | 56848 |
| Molecular Weight (g/mol) | 70 kDa |
| Name | Sphingosine Kinase 2/SPHK2 |
| Quantity | 10 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Sphingosine Kinase 2/SPHK2 protein Asn2-Pro654,with an N-terminal 5-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.