Learn More
R&D Systems™ Recombinant Human SPINK1 Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2 μm filtered solution in MES, NaCl, CaCl 2 DTT, Glycerol and Brij-35. |
| Gene ID (Entrez) | 6690 |
| Molecular Weight (g/mol) | 7 kDa |
| Name | SPINK1 |
| Quantity | 10 μg |
| Source | E. coli-derived human SPINK1 protein Asp24-Cys79,with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.