Learn More
R&D Systems™ Recombinant Human SREC-II/SCARF2 Fc Chimera Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 91179 |
| Molecular Weight (g/mol) | 69.8 kDa (monomer) |
| Name | SREC-II/SCARF2 Fc Chimera |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human SREC-II/SCARF2 protein, (N-terminus) Human SREC-II (Gln44-Gly442) Accession &Num; Q96GP6 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, -20 to -70° C under sterile conditions after opening. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.