Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Sulfamidase/SGSH Protein
SDP

Catalog No. 8380SU020 Shop All Novus Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
83-80S-U020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 83-80S-U020 Supplier R&D Systems™ Supplier No. 8380SU020

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to hydrolyze the substrate 4-Nitrocatechol Sulfate (PNCS). The specific activity is >0.3 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number P51688
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 6448
Molecular Weight (g/mol) M.W. Observed: 55-63 kDa, under reducing conditions.
Name Sulfamidase/SGSH
Quality Control Testing SDS-PAGE: 55-63kDa, reducing conditions
Quantity 20 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Sulfamidase/SGSH protein Arg23-Leu502 with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Regulatory Status RUO
Termination Arg23
Without Additives Carrier Protein
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol SGSH
Conjugate Unconjugated
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Sulfamidase/SGSH
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.