Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Sulfatase Modifying Factor 1/SUMF1 His Protein
SDP

Catalog No. NBP17254050 Shop All R&D Systems Products
Change view
Click to view available options
50μg; Unlabeled
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBP17254050 50μg; Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17254050 Supplier Novus Biologicals™ Supplier No. NBP17254050UG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 91-374 of Human Sulfatase Modifying Factor 1/SUMF1 The Recombinant Human Sulfatase Modifying Factor 1/SUMF1 Protein is derived from E. coli. The Recombinant Human Sulfatase Modifying Factor 1/SUMF1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_877437
For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl, pH8.0, 2M Urea, 20%Glycerol, 2mM DTT
Gene ID (Entrez) 285362
Molecular Weight (g/mol) 34.1kDa
Name Sulfatase Modifying Factor 1/SUMF1 Protein
Purification Method SDS-PAGE
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >85%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.