Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human TACE/ADAM17 Protein
SDP

Catalog No. 930ADB010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
930ADB010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 930ADB010 Supplier R&D Systems™ Supplier No. 930ADB010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

The Recombinant Human TACE/ADAM17 Protein is derived from Sf 21 (baculovirus). The Recombinant Human TACE/ADAM17 Protein has been validated for the following applications: Enzyme Activity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Tris, NaCl and Brij-35.
Gene ID (Entrez) 6868
Molecular Weight (g/mol) 52 kDa
Name TACE/ADAM17
Quantity 10 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human TACE/ADAM17 protein Arg215-Asn671,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Mature form. Recombinant Human TACE/ADAM17 is prone to proteolytic cleavage at C-terminus. The predominant form of the purified protein lacks the His tag.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.