Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Tau K18/Tau PHF Core Protein

Catalog No. SP496100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SP496100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SP496100 Supplier R&D Systems™ Supplier No. SP496100
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Tau K18/Tau PHF Core Protein is derived from E. coli. The Recombinant Human Tau K18/Tau PHF Core Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P10636-8
For Use With (Application) Bioactivity
Gene ID (Entrez) 4137
Molecular Weight (g/mol) 14 kDa
Quantity 100 μg
Source E. coli-derived human Tau protein Leu243-Glu372
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 24 months from date of receipt, -70 °C as supplied. 6 months, -70 °C under sterile conditions after opening.
Regulatory Status RUO
Specific Reactivity Tau
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Buffer X mg/mL (X μm) in PBS pH 7.4
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.