Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human TBCA Protein
SDP

Catalog No. NBC128769 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC128769 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC128769 Supplier Novus Biologicals™ Supplier No. NBC128769
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-108 of Human TBCA The Recombinant Human TBCA Protein is derived from E. coli. The Recombinant Human TBCA Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_004598
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 7.5), 1mM DTT, 10% glycerol
Gene ID (Entrez) 6902
Molecular Weight (g/mol) 12.8kDa
Name TBCA Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MADPRVRQIK IKTGVVKRLV KEKVMYEKEA KQQEEKIEKM RAEDGENYDI KKQAEILQES RMMIPDCQRR LEAAYLDLQR ILENEKDLEE AEEYKEARLV LDSVKLEA
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Buffer PBS (pH 7.4), 10% glycerol
Endotoxin Level <1 EU / 1 ug of protein (determined by LAL method)
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.